C12H316CEF81 Chemcia Scientific, LLC

2H-Indazole

  • MDL number MFCD20483705
  • Cas Number : 271-42-1
  • Molecular Weight : 118.13927

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5666

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5666
Availability details for
Quantity
  • Availability
  • Add to Cart