C12H316CEF83 Chemcia Scientific, LLC

(2-Amino-thiazol-4-yl)-methanol

  • MDL number MFCD08275702
  • Cas Number : 51307-43-8
  • Molecular Weight : 130.16922

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5667

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5667
Availability details for
Quantity
  • Availability
  • Add to Cart