C12H316CEF84 Chemcia Scientific, LLC

7-Trifluoromethyl-imidazo[1,2-a]pyridine-3-carboxylic acid ethyl ester

  • MDL number MFCD29037519
  • Cas Number : 1397206-76-6
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 7-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Inchi Code 1S/C11H9F3N2O2/c1-2-18-10(17)8-6-15-9-5-7(11(12,13)14)3-4-16(8)9/h3-6H,2H2,1H3
Inchi Key XIICEPYUKGVRMS-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5668

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5668
Availability details for
Quantity
  • Availability
  • Add to Cart