C12H316CEF86 Chemcia Scientific, LLC

4-Cyclopropanesulfonyl-benzoic acid methyl ester

  • MDL number MFCD29037520
  • Molecular Weight : 240.27989

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5670

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5670
Availability details for
Quantity
  • Availability
  • Add to Cart