C12H316CEF87 Chemcia Scientific, LLC

4-Cyclopropylsulfanyl-benzoic acid methyl ester

  • MDL number MFCD29037521
  • Molecular Weight : 208.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-(cyclopropylthio)benzoate
Inchi Code 1S/C11H12O2S/c1-13-11(12)8-2-4-9(5-3-8)14-10-6-7-10/h2-5,10H,6-7H2,1H3
Inchi Key ZQNVGJKWVBXDOQ-UHFFFAOYSA-N

Description

Purity 90%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5671

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5671
Availability details for
Quantity
  • Availability
  • Add to Cart