C12H316CEF89 Chemcia Scientific, LLC

5-Chloro-6-trifluoromethyl-pyridin-2-ylamine

  • MDL number MFCD16606147
  • Cas Number : 1227595-72-3
  • Molecular Weight : 196.56038

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5673

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5673
Availability details for
Quantity
  • Availability
  • Add to Cart