C12H316CEF90 Chemcia Scientific, LLC

8-Bromo-6-chloro-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD22683352
  • Cas Number : 1433822-19-5
  • Molecular Weight : 232.46791

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5686

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5686
Availability details for
Quantity
  • Availability
  • Add to Cart