C12H316CEF91 Chemcia Scientific, LLC

8-Trifluoromethyl-imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD09995161
  • Cas Number : 1018828-72-2
  • Molecular Weight : 230.1476

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5693

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5693
Availability details for
Quantity
  • Availability
  • Add to Cart