C12H316CEF93 Chemcia Scientific, LLC

1-Bromo-4-iodo-5-methoxy-2-nitro-benzene

  • MDL number MFCD29037527
  • Molecular Weight : 357.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-4-iodo-5-methoxy-2-nitrobenzene
Inchi Code 1S/C7H5BrINO3/c1-13-7-2-4(8)6(10(11)12)3-5(7)9/h2-3H,1H3
Inchi Key OHAXTKNVCPDVGV-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5700

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5700
Availability details for
Quantity
  • Availability
  • Add to Cart