C12H316CEF9B Chemcia Scientific, LLC

(3-Methoxy-2-methyl-phenyl)-methanol

  • MDL number MFCD12031988
  • Cas Number : 33797-34-1
  • Molecular Weight : 152.19479

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5716

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5716
Availability details for
Quantity
  • Availability
  • Add to Cart