C12H316CEF9D Chemcia Scientific, LLC

6-Methoxy-7-methyl-1H-[1,5]naphthyridin-4-one

  • MDL number MFCD29037533
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methoxy-7-methyl-1,5-naphthyridin-4-ol
Inchi Code 1S/C10H10N2O2/c1-6-5-7-9(12-10(6)14-2)8(13)3-4-11-7/h3-5H,1-2H3,(H,11,13)
Inchi Key KXFDRSQQOJXCIV-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5718

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5718
Availability details for
Quantity
  • Availability
  • Add to Cart