C12H316CEF9F Chemcia Scientific, LLC

8-Bromo-7-chloro-2-methoxy-3-methyl-[1,5]naphthyridine

  • MDL number MFCD29037535
  • Molecular Weight : 287.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-bromo-7-chloro-2-methoxy-3-methyl-1,5-naphthyridine
Inchi Code 1S/C10H8BrClN2O/c1-5-3-7-9(14-10(5)15-2)8(11)6(12)4-13-7/h3-4H,1-2H3
Inchi Key VSFYVHOMBLLDDR-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5720

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5720
Availability details for
Quantity
  • Availability
  • Add to Cart