C12H316CEFA4 Chemcia Scientific, LLC

(6-Chloro-2-iodo-pyridin-3-yl)-carbamic acid tert-butyl ester

  • Cas Number : 1621888-56-9
  • Molecular Weight : 354.57674

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5726

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5726
Availability details for
Quantity
  • Availability
  • Add to Cart