C12H316CEFA5 Chemcia Scientific, LLC

(4-Amino-3-chloro-phenyl)-acetic acid

  • MDL number MFCD11975979
  • Cas Number : 22106-57-6
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-amino-3-chlorophenyl)acetic acid
Inchi Code 1S/C8H8ClNO2/c9-6-3-5(4-8(11)12)1-2-7(6)10/h1-3H,4,10H2,(H,11,12)
Inchi Key RNPFHSPLVUBKAU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5733

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5733
Availability details for
Quantity
  • Availability
  • Add to Cart