C12H316CEFAF Chemcia Scientific, LLC

3-Chloro-6-methoxy-7-methyl-[1,5]naphthyridine-4-carbonitrile

  • MDL number MFCD29037540
  • Cas Number : 2065250-14-6
  • Molecular Weight : 233.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-6-methoxy-7-methyl-1,5-naphthyridine-4-carbonitrile
Inchi Code 1S/C11H8ClN3O/c1-6-3-9-10(15-11(6)16-2)7(4-13)8(12)5-14-9/h3,5H,1-2H3
Inchi Key XCEFTAMALPLWHR-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5752

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5752
Availability details for
Quantity
  • Availability
  • Add to Cart