C12H316CEFB2 Chemcia Scientific, LLC

6-Methyl-3,4-dihydro-2H-benzo[1,4]oxazine hydrochloride

  • MDL number MFCD09026861
  • Cas Number : 1095270-80-6
  • Molecular Weight : 185.65509

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Salt Data HCl
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5777

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5777
Availability details for
Quantity
  • Availability
  • Add to Cart