C12H316CEFB3 Chemcia Scientific, LLC

5-Methoxy-quinazoline-2,4-diamine

  • MDL number MFCD00665830
  • Cas Number : 27018-21-9
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methoxy-2,4-quinazolinediamine
Inchi Code 1S/C9H10N4O/c1-14-6-4-2-3-5-7(6)8(10)13-9(11)12-5/h2-4H,1H3,(H4,10,11,12,13)
Inchi Key FYEWYRWGCVGNJW-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5786

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5786
Availability details for
Quantity
  • Availability
  • Add to Cart