C12H316CEFB7 Chemcia Scientific, LLC

5-Bromo-quinazoline-2,4-diamine

  • MDL number MFCD01411817
  • Cas Number : 119584-75-7
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2,4-quinazolinediamine
Inchi Code 1S/C8H7BrN4/c9-4-2-1-3-5-6(4)7(10)13-8(11)12-5/h1-3H,(H4,10,11,12,13)
Inchi Key NPAVQJGHQXYNDS-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5801

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5801
Availability details for
Quantity
  • Availability
  • Add to Cart