C12H316CEFBC Chemcia Scientific, LLC

4-Chloro-2-chloromethyl-benzooxazole

  • MDL number MFCD24463240
  • Cas Number : 143708-34-3
  • Molecular Weight : 202.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-2-(chloromethyl)benzo[d]oxazole
Inchi Code 1S/C8H5Cl2NO/c9-4-7-11-8-5(10)2-1-3-6(8)12-7/h1-3H,4H2
Inchi Key GCQOLXQHWAPRGY-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Pink Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5841

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5841
Availability details for
Quantity
  • Availability
  • Add to Cart