C12H316CEFBD Chemcia Scientific, LLC

6-Nitro-1H-indazol-3-ylamine

  • MDL number MFCD11707246
  • Cas Number : 1027259-01-3
  • Molecular Weight : 178.15147

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Red Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5857

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5857
Availability details for
Quantity
  • Availability
  • Add to Cart