C12H316CEFBE Chemcia Scientific, LLC

7-Iodo-benzo[d]isoxazol-3-ylamine

  • MDL number MFCD29037550
  • Cas Number : 1934530-26-3
  • Molecular Weight : 260.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5C
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5861

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5861
Availability details for
Quantity
  • Availability
  • Add to Cart