C12H316CEFBF Chemcia Scientific, LLC

2,5-Dichloro-thiazolo[5,4-b]pyridine

  • MDL number MFCD18256936
  • Cas Number : 857970-66-2
  • Molecular Weight : 205.06624

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5862

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5862
Availability details for
Quantity
  • Availability
  • Add to Cart