C12H316CEFC2 Chemcia Scientific, LLC

2-Amino-4-fluoro-5-methyl-benzonitrile

  • MDL number MFCD17167315
  • Cas Number : 1037206-84-0
  • Molecular Weight : 150.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-4-fluoro-5-methylbenzonitrile
Inchi Code 1S/C8H7FN2/c1-5-2-6(4-10)8(11)3-7(5)9/h2-3H,11H2,1H3
Inchi Key YAEPXRRKVAJHHQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5871

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5871
Availability details for
Quantity
  • Availability
  • Add to Cart