C12H316CEFC8 Chemcia Scientific, LLC

2-Amino-4-fluoro-5-methyl-benzoic acid methyl ester

  • MDL number MFCD20691241
  • Cas Number : 1037206-86-2
  • Molecular Weight : 183.18395

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5883

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5883
Availability details for
Quantity
  • Availability
  • Add to Cart