C12H316CEFCE Chemcia Scientific, LLC

5-Chloro-4-fluoro-2-hydroxy-benzoic acid

  • MDL number MFCD20640444
  • Cas Number : 189283-52-1
  • Molecular Weight : 190.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-4-fluoro-2-hydroxybenzoic acid
Inchi Code 1S/C7H4ClFO3/c8-4-1-3(7(11)12)6(10)2-5(4)9/h1-2,10H,(H,11,12)
Inchi Key DDRJDFULMKUQRV-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5899

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5899
Availability details for
Quantity
  • Availability
  • Add to Cart