C12H316CEFCF Chemcia Scientific, LLC

2,4-Difluoro-5-methyl-benzonitrile

  • MDL number MFCD09835341
  • Cas Number : 329314-68-3
  • Molecular Weight : 153.13255

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5901

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5901
Availability details for
Quantity
  • Availability
  • Add to Cart