C12H316CEFD2 Chemcia Scientific, LLC

6-Chloro-5-methyl-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester

  • MDL number MFCD28099885
  • Molecular Weight : 238.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 6-chloro-5-methylimidazo[1,2-a]pyridine-2-carboxylate
Inchi Code 1S/C11H11ClN2O2/c1-3-16-11(15)9-6-14-7(2)8(12)4-5-10(14)13-9/h4-6H,3H2,1-2H3
Inchi Key KHENIFBNPRSYEY-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=5921

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5921
Availability details for
Quantity
  • Availability
  • Add to Cart