C12H316CEFD9 Chemcia Scientific, LLC

2-Amino-4-methoxy-benzothiazole-6-carboxylic acid methyl ester

  • MDL number MFCD23104009
  • Cas Number : 955886-84-7
  • Molecular Weight : 238.2668

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6033

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6033
Availability details for
Quantity
  • Availability
  • Add to Cart