C12H316CEFDD Chemcia Scientific, LLC

(5-Bromo-pyridin-3-yl)-carbamic acid ethyl ester

  • MDL number MFCD00831050
  • Cas Number : 152684-24-7
  • Molecular Weight : 245.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-bromo-3-pyridinylcarbamate
Inchi Code 1S/C8H9BrN2O2/c1-2-13-8(12)11-7-3-6(9)4-10-5-7/h3-5H,2H2,1H3,(H,11,12)
Inchi Key BFWDDTLTJVTQHZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6042

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6042
Availability details for
Quantity
  • Availability
  • Add to Cart