C12H316CEFE5 Chemcia Scientific, LLC

6-Bromo-[1,2,4]triazolo[1,5-a]pyridine-8-carboxylic acid methyl ester

  • Cas Number : 1801262-20-3
  • Molecular Weight : 256.05992

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6099

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6099
Availability details for
Quantity
  • Availability
  • Add to Cart