C12H316CEFED Chemcia Scientific, LLC

4-Amino-2-methoxy-5-nitro-benzoic acid

  • MDL number MFCD09908199
  • Cas Number : 59338-90-8
  • Molecular Weight : 212.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-amino-2-methoxy-5-nitrobenzoic acid
Inchi Code 1S/C8H8N2O5/c1-15-7-3-5(9)6(10(13)14)2-4(7)8(11)12/h2-3H,9H2,1H3,(H,11,12)
Inchi Key AOANPIGDKLHHIC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6114

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6114
Availability details for
Quantity
  • Availability
  • Add to Cart