C12H316CEFF4 Chemcia Scientific, LLC

4-Amino-5-bromo-2-methoxy-benzoic acid

  • MDL number MFCD02186027
  • Cas Number : 35290-97-2
  • Molecular Weight : 246.06186

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6121

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6121
Availability details for
Quantity
  • Availability
  • Add to Cart