C12H316CEFF5 Chemcia Scientific, LLC

2-[5-Fluoro-2-(toluene-4-sulfonylimino)-2H-pyridin-1-yl]-acetamide

  • Cas Number : 1123163-32-5
  • Molecular Weight : 323.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (Z)-2-(5-fluoro-2-(tosylimino)pyridin-1(2H)-yl)acetamide
Inchi Code 1S/C14H14FN3O3S/c1-10-2-5-12(6-3-10)22(20,21)17-14-7-4-11(15)8-18(14)9-13(16)19/h2-8H,9H2,1H3,(H2,16,19)/b17-14-
Inchi Key MKFJRJIIEZXKEU-VKAVYKQESA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin CN
Product Link http://www.chemcia.com/product_fl02.asp?id=6122

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6122
Availability details for
Quantity
  • Availability
  • Add to Cart