C12H316CEFF8 Chemcia Scientific, LLC

S-2-Amino-4-methyl-pentanoic acid dimethylamide

  • MDL number MFCD14697077
  • Cas Number : 77167-46-5
  • Molecular Weight : 158.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-2-amino-N,N,4-trimethylpentanamide
Inchi Code 1S/C8H18N2O/c1-6(2)5-7(9)8(11)10(3)4/h6-7H,5,9H2,1-4H3/t7-/m0/s1
Inchi Key VINQQWZFGIKZAZ-ZETCQYMHSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6125

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6125
Availability details for
Quantity
  • Availability
  • Add to Cart