C12H316CEFF9 Chemcia Scientific, LLC

1-Amino-cyclopropanecarboxylic acid dimethylamide

  • MDL number MFCD21238101
  • Cas Number : 1481463-48-2
  • Molecular Weight : 128.17534

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6126

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6126
Availability details for
Quantity
  • Availability
  • Add to Cart