C12H316CEFFA Chemcia Scientific, LLC

1-Amino-cyclobutanecarboxylic acid dimethylamide

  • MDL number MFCD24059179
  • Cas Number : 1508823-36-6
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-amino-N,N-dimethylcyclobutane-1-carboxamide
Inchi Code 1S/C7H14N2O/c1-9(2)6(10)7(8)4-3-5-7/h3-5,8H2,1-2H3
Inchi Key RIFVSBDIRWLMPT-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Liquid(Solid at 0oC)
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7104

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7104
Availability details for
Quantity
  • Availability
  • Add to Cart