C12H316CEFFB Chemcia Scientific, LLC

1-(3-Methoxy-2-methyl-phenyl)-ethanone

  • MDL number MFCD13689053
  • Cas Number : 24487-92-1
  • Molecular Weight : 164.20594

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6128

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6128
Availability details for
Quantity
  • Availability
  • Add to Cart