C12H316CEFFE Chemcia Scientific, LLC

3-Bromo-2H-indazole

  • MDL number MFCD27988867
  • Cas Number : 40598-94-5
  • Molecular Weight : 197.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-2H-indazole
Inchi Code 1S/C7H5BrN2/c8-7-5-3-1-2-4-6(5)9-10-7/h1-4H,(H,9,10)
Inchi Key HTKXRTUKPXEALT-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6130

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6130
Availability details for
Quantity
  • Availability
  • Add to Cart