C12H316CF001 Chemcia Scientific, LLC

(S,S)-(1-Dimethylcarbamoyl-2-methyl-butyl)-carbamic acid tert-butyl ester

  • MDL number MFCD29037636
  • Cas Number : 192821-24-2
  • Molecular Weight : 258.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl ((2S,3S)-1-(dimethylamino)-3-methyl-1-oxopentan-2-yl)carbamate
Inchi Code 1S/C13H26N2O3/c1-8-9(2)10(11(16)15(6)7)14-12(17)18-13(3,4)5/h9-10H,8H2,1-7H3,(H,14,17)/t9-,10-/m0/s1
Inchi Key MIEHGJLZGQANNB-UWVGGRQHSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6140

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6140
Availability details for
Quantity
  • Availability
  • Add to Cart