C12H316CF006 Chemcia Scientific, LLC

6-Bromo-5-chloro-1H-benzoimidazole

  • MDL number MFCD22558143
  • Cas Number : 1360953-55-4
  • Molecular Weight : 231.48033

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6148

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6148
Availability details for
Quantity
  • Availability
  • Add to Cart