C12H316CF00E Chemcia Scientific, LLC

2-Bromo-4-chloro-5-iodo-benzoic acid methyl ester

  • MDL number MFCD12547836
  • Cas Number : 1208077-68-2
  • Molecular Weight : 375.38925

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6161

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6161
Availability details for
Quantity
  • Availability
  • Add to Cart