C12H316CF014 Chemcia Scientific, LLC

1-(5-Hydroxy-2-nitro-phenyl)-ethanone

  • MDL number MFCD12827423
  • Cas Number : 30879-49-3
  • Molecular Weight : 181.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(5-hydroxy-2-nitrophenyl)ethanone
Inchi Code 1S/C8H7NO4/c1-5(10)7-4-6(11)2-3-8(7)9(12)13/h2-4,11H,1H3
Inchi Key PHQWMZYOGOPUIN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6167

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6167
Availability details for
Quantity
  • Availability
  • Add to Cart