C12H316CF017 Chemcia Scientific, LLC

2,3-Dichloro-6-trifluoromethyl-pyridine

  • MDL number MFCD02083501
  • Cas Number : 89719-90-4
  • Molecular Weight : 215.99074

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6169

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6169
Availability details for
Quantity
  • Availability
  • Add to Cart