C12H316CF019 Chemcia Scientific, LLC

4-Amino-3-bromo-2-chloro-benzoic acid

  • MDL number MFCD29067426
  • Molecular Weight : 250.4804

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6172

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6172
Availability details for
Quantity
  • Availability
  • Add to Cart