C12H316CF01D Chemcia Scientific, LLC

2-Bromo-4-methyl-5-trifluoromethyl-phenylamine

  • MDL number MFCD15527582
  • Cas Number : 1260776-57-5
  • Molecular Weight : 254.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-4-methyl-5-(trifluoromethyl)aniline
Inchi Code 1S/C8H7BrF3N/c1-4-2-6(9)7(13)3-5(4)8(10,11)12/h2-3H,13H2,1H3
Inchi Key OUBNGEOXDNRDRY-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6176

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6176
Availability details for
Quantity
  • Availability
  • Add to Cart