C12H316CF01E Chemcia Scientific, LLC

2-Iodo-4-methyl-5-trifluoromethyl-phenylamine

  • MDL number MFCD11111110
  • Cas Number : 872624-68-5
  • Molecular Weight : 301.05129

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6177

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6177
Availability details for
Quantity
  • Availability
  • Add to Cart