C12H316CF020 Chemcia Scientific, LLC

5-Chloro-4-iodo-2-methoxy-benzoic acid

  • MDL number MFCD09738653
  • Cas Number : 473264-13-0
  • Molecular Weight : 312.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-4-iodo-2-methoxybenzoic acid
Inchi Code 1S/C8H6ClIO3/c1-13-7-3-6(10)5(9)2-4(7)8(11)12/h2-3H,1H3,(H,11,12)
Inchi Key LLLOGZBHLFGNSY-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Red Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6179

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6179
Availability details for
Quantity
  • Availability
  • Add to Cart