C12H316CF02A Chemcia Scientific, LLC

N-(3-Hydroxy-4-methyl-phenyl)-2,2-dimethyl-propionamide

  • MDL number MFCD20037146
  • Cas Number : 1312305-91-1
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(3-hydroxy-4-methylphenyl)-2,2-dimethylpropanamide
Inchi Code 1S/C12H17NO2/c1-8-5-6-9(7-10(8)14)13-11(15)12(2,3)4/h5-7,14H,1-4H3,(H,13,15)
Inchi Key ABOSOXSRKQXGLC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6188

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6188
Availability details for
Quantity
  • Availability
  • Add to Cart