C12H316CF02B Chemcia Scientific, LLC

(6-Trifluoromethyl-pyridin-3-yl)-hydrazine

  • MDL number MFCD11113388
  • Cas Number : 1035173-53-5
  • Molecular Weight : 177.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-hydrazino-2-(trifluoromethyl)pyridine
Inchi Code 1S/C6H6F3N3/c7-6(8,9)5-2-1-4(12-10)3-11-5/h1-3,12H,10H2
Inchi Key KLLXIDZXMUALHC-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6189

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6189
Availability details for
Quantity
  • Availability
  • Add to Cart