C12H316CF02D Chemcia Scientific, LLC

2-Hydroxy-5-iodo-4-methoxy-benzaldehyde

  • MDL number MFCD16249658
  • Cas Number : 237056-75-6
  • Molecular Weight : 278.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-hydroxy-5-iodo-4-methoxybenzaldehyde
Inchi Code 1S/C8H7IO3/c1-12-8-3-7(11)5(4-10)2-6(8)9/h2-4,11H,1H3
Inchi Key YPUMFVXSTKNWRE-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=6191

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6191
Availability details for
Quantity
  • Availability
  • Add to Cart